Learn More
Medchemexpress LLC SRX3177 | 2241237-51-2 | C31H32N6O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15389410MM/1ML
Encompass_Preferred
SRX3177 is a triple inhibitor of CDK4/6, PI3K, and BRD4. This compound blocks the interaction between the SARS-CoV-2 E protein and the BRD2/4 BD1 domain, which helps restore E protein-attenuated NF-κB activity. SRX3177 shows broad cytotoxic activity against cancer cells and can be used for the study of anti-SARS-CoV-2 and cancer.
  • Inhibits CDK4/6, PI3K, and BRD4
  • Blocks interaction between SARS-CoV-2 E protein and BRD2/4 BD1 domain
  • Restores E protein-attenuated NF-κB activity
  • Exhibits broad cytotoxic activity against cancer cells
  • Useful for anti-SARS-CoV-2 research
  • Useful for cancer research
Catalog No. 50-003-76337