Learn More
TARGETMOL CHEMICALS INC SRX3207 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T224305MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. SRX3207 is an inhibitor of Syk and PI3K and relieves tumor immunosuppression. purity: 98%
Catalog No. 50-258-0955