Learn More
Grainger CAFRAMO Dispersion Blade, Stainless
Supplier:  Grainger 38C850
Encompass_Preferred
The CAFRAMO Dispersion Blade is a stainless steel dispersion blade designed for use with overhead stirrers. It features a propeller design with 3 blades and is made of 316 stainless steel for corrosion resistance. This dispersion blade has a shaft diameter of 10mm and an overall blade diameter of 50mm. It is rated for speeds up to 3000 RPM. The CAFRAMO dispersion blade is suitable for homogenizing, dispersing, emulsifying, and mixing applications with viscosities up to 2000 mPas. This high shear mixing blade generates vigorous circulation and vortex formation for fast, uniform blending.
Catalog No. NC2828006