Learn More
New Jersey Fluid Systems LLC SS-12-RB-4, SS PIPE FITTING
Supplier:  New Jersey Fluid Systems LLC SS12RB4
Encompass
Stainless Steel Pipe Fitting, Reducing Bushing, 3/4 in. Male NPT x 1/4 in. Female NPT
Catalog No. NC2067441