Learn More
BOSTON FLUID SYSTEMS INC SS 1PC 40G SRS 3-WAY BALL VLV
Supplier:  BOSTON FLUID SYSTEMS INC SS43GXS41466
Encompass
Stainless Steel 1-Piece 40G Series 3-Way Ball Valve, 0.90 Cv, 1/4 in. Swagelok Tube Fitting, No Lube, SC-11 Cleaned
Catalog No. NC2949898