Learn More
BOSTON FLUID SYSTEMS INC Stainless Steel 1-Piece 40G Series Ball Valve
Supplier:  BOSTON FLUID SYSTEMS INC SS43GS4
Encompass
Stainless Steel 1-Piece 40G Series Ball Valve, 1.4 Cv, 1/4 in. Swagelok Tube Fitting.
Catalog No. NC1411377