Learn More
Strem, An Ascensus Company CAS 29841-69-8. 500mg. (1S2S)-(-)-12-Diphenylethylenediamine min. 97% (SS)-DPEN. MFCD00082751
SDP
Supplier:  Strem, An Ascensus Company 070475500MG
Encompass
CAS 29841-69-8. 500mg. (1S2S)-(-)-12-Diphenylethylenediamine min. 97% (SS)-DPEN. MFCD00082751. Molecular Weight 212.29. Molecular Formula (C6H5)(NH2)CHCH(NH2)(C6H5). Color/form white to pale yellow xtl. Strem 07-0475. http//www.strem.com/catalog/v/07-0475/
Catalog No. 50-211-0696