Learn More
BOSTON FLUID SYSTEMS INC SS MINI QCK CONNECT BDY 1/16IN
Supplier:  BOSTON FLUID SYSTEMS INC SSQM2B100
Encompass
Stainless Steel Miniature Quick Connect Body, 0.05 Cv, 1/16 in. Swagelok Tube Fitting
Catalog No. NC2891497