Learn More
BOSTON FLUID SYSTEMS INC SS MINI QCK CONNECT STM W VLV
Supplier:  BOSTON FLUID SYSTEMS INC SSQM2D100
Encompass
Stainless Steel Miniature Quick Connect Stem with Valve, 0.05 Cv, 1/16 in. Swagelok Tube Fitting
Catalog No. NC2891502