Learn More
BOSTON FLUID SYSTEMS INC SS MINI QCK CONNECT STM WO VLV
Supplier:  BOSTON FLUID SYSTEMS INC SSQM2S100
Encompass
Stainless Steel Miniature Quick Connect Stem without Valve, 0.06 Cv, 1/16 in. Swagelok Tube Fitting
Catalog No. NC2891506