Learn More
Strem, An Ascensus Company CAS 71042-54-1. 250mg. (2S3S)-()-23-Bis(diphenylphosphino)-bicyclo[2.2.1]hept-5-ene min. 95% (SS)-NORPHOS. MFCD00085364
SDP
Supplier:  Strem, An Ascensus Company 150141250MG
Encompass
CAS 71042-54-1. 250mg. (2S3S)-()-23-Bis(diphenylphosphino)-bicyclo[2.2.1]hept-5-ene min. 95% (SS)-NORPHOS. MFCD00085364. Molecular Weight 462.51. Molecular Formula C31H28P2. Color/form white xtl. Strem 15-0141. http//www.strem.com/catalog/v/15-0141/
Catalog No. 50-211-0729