Learn More
Inter Dyne Systems Inc SS PEG BOARD 24X30 32PEGSSS
Supplier:  Inter Dyne Systems Inc V2430
Encompass_Preferred
Mod-Rack stainless steel pegboard 24 in. w x 30 in. h w integral drip trough, drain tubing, 32 white 6 in. long polyproplyne pegs and a wall mounting bracket.
Catalog No. NC9038328