Learn More
New Jersey Fluid Systems LLC SS PIPE FIT 3/8 MALE NPT X 1/4
Supplier:  New Jersey Fluid Systems LLC SS6RB4
Encompass
316 Stainless Steel Swagelok Pipe Reducing Bushing. 3/8 in. male NPT x 1/4 in. Female NPT
Catalog No. NC0727008