Learn More
Electron Microscopy Sciences Stainless Steel Rodent Brain Matrix Large Rat, Sagital, 300-600g
SDP
Supplier:  Electron Microscopy Sciences 69092S
Encompass_Preferred
Stainless steel matrices with 0.5 mm channel for slicing rodent brain, heart, and similar organs as thin as possible. Made of high-quality stainless steel. They can be repeatedly used, cleaned, autoclaved, and frozen. Produce fine slices. Accurate sectioning. Easy cleaning and easy handling.
Type: Sagital
Size: 300-600g
Catalog No. 50-339-03