Learn More
Cole-Parmer Cole-Parmer 3"-Diameter Sieve, Full Height Stainless Steel, No. 20 (850µ)
Supplier:  Cole-Parmer 5998711
Encompass_Preferred
Made of stainless steel; 3" diameter, full height sieve, 1" stacking height.
Catalog No. NC1124165