Learn More
New Jersey Fluid Systems LLC SS TFTG, REDUCER, 1/4IN T OD
Supplier:  New Jersey Fluid Systems LLC SS400R12
Encompass
Stainless Steel Swagelok Tube Fitting, Reducer, 1/4 in. Tube OD x 3/4 in. Swagelok Tube Adapter
Catalog No. NC2152806