Learn More
Perkin Elmer US LLC Stainless Steel Tenax GR 60/80 Packed ATD Sample Tubes, Pkg. 10
Supplier:  Perkin Elmer US LLC N9307004
Encompass_Preferred
The Tenax GR is conditioned at 320°C. PerkinElmer recommends a maximum temperature of 350°C for this sample tube with a desorption temperature of 300°C. The approximate analyte volatility range is n-C7 to n-C26 and the typical analytes for the Tenax GR are Alkyl benzenes, PAH, PCBs, which is the same as the Tenax TA. Compatible for pumped monitoring and suitable for direct sampling of solids and liquids. PerkinElmer tubes are manufactured to the tightest specifications to ensure trouble free operation and high quality analysis results. Use with PTFE Caps and O-Rings (N6200119).Tubes can also be purchased without packing (M0413595).
Catalog No. 50-905-2371