Learn More
Microsolv Technology Corporation Ss Tubing Precut .005X50Cm Red
Supplier:  Microsolv Technology Corporation 4940550
Encompass_Preferred
Tubing, Stainless Steel, Pre-Cut, 0.005 ID, 1/16 OD, HPLC Grade, Red Band, 50cm Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51667