Learn More
Aviva Systems Biology Corp SSBP2 Antibody - N-terminal region (ARP31828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSBP2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SSBP2. Alternate names: DKFZp686F03273, HSPC116, SOSS-B2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54331