Learn More
Sigma Aldrich Fine Chemicals Biosciences SSC Buffer 20x for Norther
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 9301710LF
Encompass
SSC Buffer 20x Concentrate is a standard reagent in Southern and Northern blotting procedures. This concentrated formula can be used straight or diluted to make SSC working SOLUTIONons with concentrations as low as 0.5x.
Catalog No. 50-290-7391