Learn More
Alkali Scientific SSC Buffer 20X Powder, 263.6 Grams of Powder, Makes 1 Liter of 20X Solution
SDP
Supplier:  Alkali Scientific SSC001CP
Encompass_Preferred
SSC Buffer 20X Powder (263.6 Grams) is a concentrated solution commonly used in DNA hybridization experiments. The powder form allows labs to prepare the solution in the exact volume needed by dissolving it in distilled water. This 20X concentrated solution is widely used in molecular biology for nucleic acid hybridization, particularly in Southern and Northern blotting, where high salt concentrations help in stabilizing the probe-target interactions. The SSC buffer helps maintain an optimal environment for hybridization, ensuring that DNA or RNA strands bind specifically to their complementary sequences during experimentation. This 1-liter solution is critical for efficient and accurate nucleic acid analysis.
Catalog No. 50-006-61599