Learn More
ABclonal Technology SSC4D Rabbit pAb
SDP
Supplier:  ABclonal Technology A159571000UL
Encompass_Preferred
The scavenger receptor cysteine-rich (SRCR) superfamily is an ancient and highly conserved group of cell surface and/or secreted proteins, some of which are involved in the development of the immune system and the regulation of both innate and adaptive immune responses. Group B SRCR domains usually contain 8 regularly spaced cysteines that give rise to a well-defined intradomain disulfide-bond pattern.
Catalog No. 50-001-07519