Learn More
Selleck Chemical LLC ssDNA salmon sperm E4505-1G
SDP
Supplier:  Selleck Chemical LLC E45051G
Encompass_Preferred
Deoxyribonucleic acid single-stranded from salmon testes is a ready-to-use solution of a high-quality sonicated single-stranded template nonspecific DNA blocking agent that reduces background in nucleic acid hybridization
Catalog No. 50-000-39601