Learn More
MSC SSI Schaefer Storage Tote, Polypropylene, 44 lb Capacity, Red, Stacking, Nesting
Supplier:  MSC 09053208
Encompass
This storage tote is constructed from polypropylene and has a 44 lb capacity. It features both stacking and nesting capabilities, making it ideal for efficient storage and organization. The tote is red and has a volume capacity of 2.01 cubic feet. It is also FDA approved, ensuring suitability for various applications.
  • Polypropylene construction
  • 44 lb load capacity
  • Stacking design for vertical storage
  • Nesting design for compact storage when empty
  • Red color for easy identification
  • 2.01 cu. ft. volume capacity
  • FDA approved
  • Manufactured in Germany
Catalog No. 50-005-29474