Learn More
Medchemexpress LLC HY-138936 5mg , SSK1 CAS:2629250-69-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1389365MG
Encompass_Preferred
HY-138936 5mg SSK1 CAS: 2629250-69-5 SSK1, a senescence-specific killing compound, is a β-galactosidase-targeted prodrug attenuates inflammation. SSK1 is activated by lysosomal β-galactosidase and selectively killed senescent cells through the activation of p38 MAPK and induction of apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1434