Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SsK36 TFA | 99.3% | 1771.04 (free base) | 5 MG
SDP

Supplier:  Medchemexpress LLC HYP10463A5MG

Encompass_Preferred

ssK36 TFA is a supersubstrate peptide of the histone methyltransferase (SET) domain protein 2 (SETD2). It is designed for the SETD2 protein, a specific PKMT, responsible in human cells for adding methyl groups to the 36th lysine residue of histone H3 (H3K36) to form H3K36me3. This peptide can be methylated by SETD2 at a rate more than 100 times faster than the natural substrate H3K36 and can be used to study the catalytic mechanism of PKMTs, especially substrate specificity and catalytic efficiency.

  • Purity of 99.34%
  • Molecular weight of 1771.04 (free base)
  • Formula: C79H127N29O18.xC2HF3O2
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Ala-Pro-Arg-Phe-Gly-Gly-Val-Lys-Arg-Pro-Asn-Arg-Tyr-Arg-Pro
  • Sequence shortening: APRFGGVKRPNRYRP
  • Solubility (in vitro): 1 mg/mL in H2O (requires ultrasonic)

Catalog No. 50-004-24544


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.