Learn More
Chemglass Life Sciences 2-Port Assembly, 1000mL Spinner Flask
SDP
Supplier:  Chemglass Life Sciences CLS142702
Encompass
Port assemblies are constructed of 316L electro-polished stainless steel.These port assemblies fit all types of spinner flasks.The assemblies consist of a siphon and vent tube.Supplied complete with a GL-45 open top cap and silicone gasket.
Catalog No. 50-121-4588