Learn More
Bellco Glass, Inc. Stainless Steel 2-Port Assembly , 3 L For Angled Sidearms
SDP
Supplier:  Bellco Glass, Inc. 196520003R
Encompass_Preferred
All port assemblies are manufactured from 316l stainless steel and have an electropolish finish. 2- port assemblies have siphon and vent tubes. 3-port assemblies have siphon, fill tube and vent tubes. All port assemblies include a 45mm blue polypropylene cap. MADE IN THE USA.
Catalog No. 50-194-4990