Learn More
Bellco Glass, Inc. Stainless Steel 3-Port Assembly , 500mL For Angled 45mm Sidearms
SDP
Supplier:  Bellco Glass, Inc. 196530005R
Encompass_Preferred
Seam welded with electropolished finish3-port with siphon, fill and vent tubesIncludes 45mm pp cap and silicone gasket500ml with 100mm cap Longest tube: 1.5". MADE IN THE USA.
Catalog No. 50-194-5004