Learn More
Bellco Glass, Inc. Stainless Steel Bent Harvesting Tube,5L 1/2 OD x 16"- Flat Bottom Vessel"
SDP
Supplier:  Bellco Glass, Inc. 196491105
Encompass_Preferred
316l stainless steel bent harvesting tube for 5l flat bottom omni vessel. Use with large flange head-plate. MADE IN THE USA. Size: 1/2" x 16" Id: .430"
Catalog No. 50-194-4941