Learn More
Bellco Glass, Inc. Stainless Steel Fitting w/Cap,12mm
SDP
Supplier:  Bellco Glass, Inc. 196521060
Encompass_Preferred
Probe adapters & compression fittings. Bellco compression fittings are designed using our standard 45 mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and do monitoring via insertion of these probes into the sidearms of vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200 mm stainless steel headplate or a 45 mm vertical sidearm. MADE IN THE USA. Probe OD: 12mm Stainless steel fitting w/ cap 12mm
Catalog No. 50-194-4999