Learn More
Bellco Glass, Inc. Stainless Steel Sparge Assembly w/Frt, 3/8 For 3L Bioreactor System
SDP
Supplier:  Bellco Glass, Inc. 196402040
Encompass_Preferred
316l stainless steel sparger assembly with stainless steel frit for 3 liter bioreactor system. MADE IN THE USA English threads Material: 316l SST with electropolish finish ra 32 or better tubing specifications: .250" od x .152" idOverall length: 10 43/64"Inseration Length: 9 1/4" With SST Frit - 10µ Supply with SST Nut & O-ring
Catalog No. 50-194-4722