Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack, 16mm-24P 4x6 Spacing
SDP
Supplier:  Bellco Glass, Inc. 202716024
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 11/16" (17.5mm) Tube size: 16mm Spacing: 4x6 holds 24 tubes Dimensions: 4 3/4" x 3 1/4" x 3" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5303