Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack,38mm-12P 2x6 Spacing
SDP
Supplier:  Bellco Glass, Inc. 202738206
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 1 9/16" (39.6mm) Tube size: 38mm Spacing: 2x6 holds 12 tubes Dimensions: 10" x 3 1/2" x 2 7/8" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .162"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5324