Learn More
Medchemexpress LLC SSTC3 | 1242422-09-8 | 99.4% | 1 ML

Supplier: Medchemexpress LLC HY12067510MM 1ML
SSTC3 is a small-molecule activator of casein kinase 1α (CK1α) that inhibits WNT signaling with nanomolar potency. It is supplied as a high-purity solid and as a 10 mM solution in DMSO for research use.
- Activates CK1α with Kd ≈ 32 nM and inhibits WNT signaling (EC50 ≈ 30 nM).
- High reported purity (≈99.4%), suitable for biochemical studies.
- Supplied as 10 mM in DMSO (1 mL) or as solid quantities for in vitro research.
- Soluble in DMSO (125 mg/mL) and compatible with common in vivo formulations at ≥2.08 mg/mL.
- Stable when stored as powder at -20°C (up to 3 years) and in solution at -80°C (up to 2 years).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.