Learn More
Medchemexpress LLC HY-101441 5mg Medchemexpress, ST-193 CAS:489416-12-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1014415MG
Encompass_Preferred
Medchemexpress, HY-101441 5mg ST-193 CAS:489416-12-8 ST-193 is a potent broad-spectrum arenavirus inhibitor; inhibits Guanarito, Junin, Lassa and Machupo virus with IC 50 values of 0.44, 0.62, 1.4 and 3.1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2130