Learn More
Bellco Glass, Inc. Straight Harvesting Pipe Assembly , 15L 10mm Port
SDP
Supplier:  Bellco Glass, Inc. 790915113
Encompass_Preferred
Used for withdraw of contents from 3L bioreactor vessel. Fits 10mm port on 3L metric headplate. MADE IN THE USA
Catalog No. 50-194-6494