Learn More
Medchemexpress LLC ST6GALNAC2 Human H 10ug
SDP
Supplier:  Medchemexpress LLC HYP7133210UG
Encompass_Preferred
ST6GALNAC2 Protein, human (HEK293, His) is a recombinant sialyltransferase supplied as a ready-to-use, frozen solution for biochemical and glycosylation studies. The preparation is formulated to preserve activity and stability, with low endotoxin suitable for in vitro assays, and is accompanied by a manufacturer datasheet documenting activity and storage recommendations.
  • Recombinant human sialyltransferase expressed in HEK293 cells.
  • His-tagged construct for detection and purification.
  • Formulated in buffered solution with trehalose for stability.
  • Low endotoxin suitable for most in vitro applications.
  • Validated specific activity for enzymatic assays.
  • Shipped on dry ice; recommended storage at -80°C.
Catalog No. 50-001-96642