Learn More
STA PHARMACEUTICAL US LLC methyltetrazine-NHS ester | 5 g | CAS 1644644-96-1 | InChIKey BIHJLZOOHNOUCG-UHFFFAOYSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAA122665GM
methyltetrazine-NHS ester is a Amino Acid reagent (Subcategory: Building Block) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1644644-96-1
- MDL: No data
- InChIKey: BIHJLZOOHNOUCG-UHFFFAOYSA-N
- Molecular Weight: 327.3
- Molecular Formula: C15H13N5O4
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: 2,5-dioxopyrrolidin-1-yl 2-(4-(6-methyl-1,2,4,5-tetrazin-3-yl)phenyl)acetate
- SMILES: CC1=NN=C(C2=CC=C(C=C2)CC(ON3C(CCC3=O)=O)=O)N=N1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.