Learn More
STA PHARMACEUTICAL US LLC PMO A(Bz) Monomer | 1 g | CAS 956139-18-7 | InChIKey HRNHOEBQNIMEGD-RJFDGRJRSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10572KA601GM
PMO A(Bz) Monomer is a Amidite reagent (Subcategory: PMO Monomer) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 956139-18-7
- MDL: No data
- InChIKey: HRNHOEBQNIMEGD-RJFDGRJRSA-N
- Molecular Weight: 722.182761998
- Molecular Formula: C38H37ClN7O4P
- Purity: ≥99%
- Container Type: 60 mL Glass (20-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 63 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: ((2S,6R)-6-(6-benzamido-9H-purin-9-yl)-4-tritylmorpholin-2-yl)methyl dimethylphosphoramidochloridate
- SMILES: O=P(Cl)(OC[C@H]1O[C@H](CN(C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)N5C6=C(N=C5)C(NC(C7=CC=CC=C7)=O)=NC=N6)N(C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.