Learn More
STA PHARMACEUTICAL US LLC 1-DMTr-2-propanediyl amidite | 25 g | InChIKey XCLJWHBPKOARSH-UHFFFAOYSA-N | MW 578.689761998
Supplier: STA PHARMACEUTICAL US LLC STAAM1092225GM
1-DMTr-2-propanediyl amidite is a Amidite reagent (Subcategory: Spacer Modifiers) sold by WuXi TIDES. Offered in 25 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: No data
- MDL: No data
- InChIKey: XCLJWHBPKOARSH-UHFFFAOYSA-N
- Molecular Weight: 578.689761998
- Molecular Formula: C33H43N2O5P
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: 1-(bis(4-methoxyphenyl)(phenyl)methoxy)propan-2-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(OP(OCCC#N)N(C(C)C)C(C)C)COC(C1=CC=CC=C1)(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.