Learn More
Strem, An Ascensus Company CAS# 56553-60-7. 100G. Sodium triacetoxyborohydride, min. 95%. MFCD00012211
SDP
Supplier:  Strem, An Ascensus Company 110595100G
Encompass
CAS# 56553-60-7. 100G. Sodium triacetoxyborohydride, min. 95%. MFCD00012211. Molecular Formula: Na(OOCCH3)3BH. Molecular Weight: 211.94. Color/Form: white pwdr. Stability: air sensitive, moisture sensitive, store cold (18°C). Strem# 11-0595. www.strem.com/catalog/v3/11-0595/
Catalog No. 50-196-3583