Learn More
Hach Company Stablcal Turbidity Standard, 200 NTU, 100 mL
Supplier:  Hach Company 2660442
Encompass
Stablcal Turbidity Standard for all Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 200. Bottle: 100 mL. For TL23, TU52, 2100Q, 2100N, 2100AN series.
Catalog No. 50-199-6491