Learn More
Medchemexpress LLC HY-N0738 100mg Medchemexpress, Stachydrine hydrochloride CAS:4136-37-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0738100MG
Encompass_Preferred
Medchemexpress, HY-N0738 100mg Stachydrine hydrochloride CAS:4136-37-2 Stachydrine hydrochloride is the major active constituent of Herba Leonuri, which is a potential therapy for cardiovascular diseases [2] . Stachydrine can inhibit the NF-κB signal pathway. Anti-hypertrophic activities [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3695