Learn More
MSC Akro-Mils Stacking Bin, High-Density Polypropylene, 10-7/8 in. x 17-1/2 in. x 12-1/2 in., 1 ea.
Supplier:  MSC 74124496
Encompass
This heavy-duty industrial hopper stacking bin is designed for efficient inventory management and organized parts handling. Molded from high-density polypropylene, this one-piece construction is water, rust, and corrosion-proof, and is capable of withstanding weak acids and alkalis. Its sturdy design is suitable for use on floors or standard industrial shelving to help minimize parts handling and shorten assembly times.
  • Features a load capacity of 75 lbs.
  • Stackable design includes an anti-slide stop to prevent shifting.
  • Curved-bottom hopper front facilitates easy retrieval of small parts.
  • Reinforced side ribs provide strength and prevent spreading under heavy loads.
  • Large front label slot supports efficient inventory identification.
  • Autoclavable up to 250° F.
  • Includes a large comfort grip handle for portability.
Catalog No. 50-005-13063