Learn More
Analytical Sales & Services STACKABLE VIAL RACK
Supplier:  Analytical Sales & Services 90002
Encompass
Stackable Vial Rack/Insert Tray, 25 Position for 15mm Vials
Catalog No. NC2873473
Only null left
Ships from supplier