Learn More
Meiji Techno America Meiji Techno America Stage micrometer, 1mm divided into 100 units, 0.01mm
Supplier:  Meiji Techno America MA285
Encompass_Preferred
Stage micrometer, 1mm divided into 100 units, 0.01mm
Catalog No. NC9167561