Learn More
KLARMANN RULINGS INC Stage Micrometer 1 mm, 100 Divisions
Supplier:  KLARMANN RULINGS INC KR851
Encompass_Preferred
1mm Stage micrometer with 100 divisions. Each division = 0.01mm per division. Numbered 0.0 thru 1.0. The locating circle can be removed.
Catalog No. NC9442550