Learn More
Electron Microscopy Sciences EMS Microscope Stage Micrometer SM-4
SDP
Supplier:  Electron Microscopy Sciences 602104PGC
Encompass_Preferred
Chrome on Glass. Linear Reference Scales: Glass Size - 1" x 3". Scale Length - 1 inch X-axis only, 25 mm X-axis only, Divisions- English = 0.0005”; Metric = 10um (0.010 mm), With numerical labeling every 0.1 mm and 0.5”. All EMS micrometers come in a sturdy wooden box. With Calibration.
Catalog No. 50-340-19