Learn More
Ethos Biosciences Natt-Herricks Solution, 16oz, Ethos
Supplier:  Ethos Biosciences 559316
Encompass_Preferred
Natt–Herrick’s Solution is a staining and diluting reagent designed for hematology applications. Commonly used for manual counting of blood cells, it enables simultaneous staining and dilution of specimens for improved visibility and accurate enumeration under the microscope. The formulation provides clear contrast of cellular components, supporting reliable red and white blood cell counts in manual hemocytometer procedures. Ideal for routine hematology workflows and research applications, Natt–Herrick’s Solution delivers consistent and reproducible results.
Catalog No. 50-005-74287